Assay Details
Gene Symbol: Ttll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021258836.1 | XP_021114495.1 | 5 - 6 | 791 | 61 | |
XM_021258837.1 | XP_021114496.1 | 6 - 7 | 907 | 61 | ||
XM_021258838.1 | XP_021114497.1 | 5 - 6 | 1535 | 61 | ||
XM_021258839.1 | XP_021114498.1 | 6 - 7 | 893 | 61 | ||
XM_021258840.1 | XP_021114499.1 | 6 - 7 | 930 | 61 | ||
XM_021258841.1 | XP_021114500.1 | 5 - 6 | 830 | 61 | ||
XM_021258842.1 | XP_021114501.1 | 5 - 6 | 1521 | 61 | ||
XM_021258843.1 | XP_021114502.1 | 5 - 6 | 731 | 61 | ||
XM_021258844.1 | XP_021114503.1 | 5 - 6 | 625 | 61 | ||
XM_021258845.1 | XP_021114504.1 | 5 - 6 | 770 | 61 | ||
XM_021258846.1 | XP_021114505.1 | 5 - 6 | 791 | 61 | ||
XM_021258847.1 | XP_021114506.1 | 5 - 6 | 791 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
