Assay Details
Gene Symbol: CUNH3orf33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004834495.2 | XP_004834552.1 | 4 - 5 | 545 | 101 | |
XM_021253422.1 | XP_021109081.1 | 4 - 5 | 468 | 101 | ||
XM_021253426.1 | XP_021109085.1 | 4 - 5 | 834 | 101 | ||
XM_021253429.1 | XP_021109088.1 | 4 - 5 | 379 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
