Assay Details
Gene Symbol: Lnx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004849713.2 | XP_004849770.1 | 6 - 7 | 1648 | 67 | |
XM_004849714.2 | XP_004849771.1 | 5 - 6 | 1499 | 67 | ||
XM_021262339.1 | XP_021117998.1 | 9 - 10 | 1959 | 67 | ||
XM_021262340.1 | XP_021117999.1 | 8 - 9 | 1824 | 67 | ||
XM_021262341.1 | XP_021118000.1 | 7 - 8 | 1773 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
