Assay Details
| Gene Symbol: CUNH11orf74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852011.3 | XP_004852068.1 | 3 - 4 | 353 | 85 | |
| XM_004852012.3 | XP_004852069.1 | 2 - 3 | 249 | 85 | ||
| XM_021264333.1 | XP_021119992.1 | 3 - 4 | 280 | 85 | ||
| XM_021264334.1 | XP_021119993.1 | 3 - 4 | 244 | 85 | ||
 Back To Top
 Back To Top
 CUNH11orf74
CUNH11orf74 Genomic Map
Genomic Map Back To Top
 Back To Top Amplicon length between 71 and 85
							Amplicon length between 71 and 85