Assay Details
Gene Symbol: CUNH11orf74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004852011.3 | XP_004852068.1 | 4 - 5 | 516 | 117 | |
XM_004852012.3 | XP_004852069.1 | 3 - 4 | 412 | 117 | ||
XM_021264333.1 | XP_021119992.1 | 4 - 5 | 443 | 117 | ||
XM_021264334.1 | XP_021119993.1 | 4 - 5 | 407 | 117 |

More Information
Set Membership: |
![]() ![]() |
