Assay Details
Gene Symbol: Stk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004850156.3 | XP_004850213.1 | 9 - 10 | 1291 | 94 | |
XM_021262682.1 | XP_021118341.1 | 9 - 10 | 1291 | 94 | ||
XM_021262683.1 | XP_021118342.1 | 7 - 8 | 958 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
