Assay Details
Gene Symbol: Znf608 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004854467.3 | XP_004854524.1 | 3 - 4 | 2012 | 59 | |
XM_004854468.3 | XP_004854525.1 | 3 - 4 | 1586 | 59 | ||
XM_004854470.3 | XP_004854527.1 | 2 - 3 | 495 | 59 | ||
XM_021266260.1 | XP_021121919.1 | 2 - 3 | 1152 | 59 | ||
XM_021266261.1 | XP_021121920.1 | 2 - 3 | 296 | 59 | ||
XM_021266262.1 | XP_021121921.1 | 2 - 3 | 379 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
