Assay Details
Gene Symbol: Pcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004865347.3 | XP_004865404.1 | 3 - 4 | 376 | 62 | |
XM_004865348.3 | XP_004865405.1 | 3 - 4 | 311 | 62 | ||
XM_021245259.1 | XP_021100918.1 | 5 - 6 | 678 | 62 | ||
XM_021245260.1 | XP_021100919.1 | 5 - 6 | 676 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
