Assay Details
| Gene Symbol: Maco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850610.2 | XP_004850667.1 | 8 - 9 | 1494 | 67 | |
| XM_021263084.1 | XP_021118743.1 | 8 - 9 | 1717 | 67 | ||
More Information
Set Membership: |
|

Maco1
Genomic Map