Assay Details
| Gene Symbol: Snrpb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840845.2 | XP_004840902.1 | 6 - 7 | 758 | 80 | |
| XM_004840848.3 | XP_004840905.1 | 6 - 7 | 675 | 80 | ||
| XM_021255095.1 | XP_021110754.1 | 6 - 7 | 773 | 80 | ||
| XM_021255096.1 | XP_021110755.1 | 6 - 7 | 735 | 80 | ||
| XM_021255097.1 | XP_021110756.1 | 6 - 7 | 592 | 80 | ||
More Information
Set Membership: |
|

Snrpb2
Genomic Map