Assay Details
| Gene Symbol: Narf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861042.3 | XP_004861099.1 | 7 - 8 | 938 | 61 | |
| XM_013078204.2 | XP_012933658.1 | 8 - 9 | 942 | 61 | ||
| XM_021241127.1 | XP_021096786.1 | 9 - 10 | 988 | 61 | ||
| XM_021241128.1 | XP_021096787.1 | 8 - 9 | 962 | 61 | ||
| XM_021241129.1 | XP_021096788.1 | 8 - 9 | 1008 | 61 | ||
More Information
Set Membership: |
|

Narf
Genomic Map