Assay Details
| Gene Symbol: Pxmp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867929.3 | XP_004867986.1 | 3 - 4 | 452 | 80 | |
| XM_004867930.3 | XP_004867987.1 | 3 - 4 | 442 | 80 | ||
| XM_004867931.3 | XP_004867988.1 | 2 - 3 | 392 | 80 | ||
More Information
Set Membership: |
|

Pxmp4
Genomic Map