Assay Details
| Gene Symbol: Dtnbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847653.3 | XP_004847710.1 | 3 - 4 | 279 | 76 | |
| XM_004847656.3 | XP_004847713.1 | 3 - 4 | 289 | 76 | ||
| XM_004847657.3 | XP_004847714.1 | 3 - 4 | 323 | 76 | ||
| XM_021260622.1 | XP_021116281.1 | 3 - 4 | 267 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021260622.1 transcripts.
Set Membership: |
|

Dtnbp1
Genomic Map