Assay Details
| Gene Symbol: Duoxa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835477.3 | XP_004835534.2 | 6 - 7 | 990 | 56 | |
| XM_021264801.1 | XP_021120460.1 | 5 - 6 | 932 | 56 | ||
More Information
Set Membership: |
|

Duoxa1
Genomic Map