Assay Details
| Gene Symbol: Dhfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841619.2 | XP_004841676.1 | 4 - 5 | 456 | 71 | |
| XM_013071084.2 | XP_012926538.1 | 4 - 5 | 423 | 71 | ||
| XM_013071085.2 | XP_012926539.1 | 4 - 5 | 399 | 71 | ||
| XM_013071086.2 | XP_012926540.1 | 4 - 5 | 303 | 71 | ||
| XM_013071087.2 | XP_012926541.2 | 4 - 5 | 555 | 71 | ||
| XM_021255715.1 | XP_021111374.1 | 4 - 5 | 573 | 71 | ||
More Information
Set Membership: |
|

Dhfr
Genomic Map