Assay Details
Gene Symbol: Dyrk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004862060.3 | XP_004862117.1 | 2 - 3 | 1048 | 70 | |
XM_004862061.3 | XP_004862118.1 | 2 - 3 | 425 | 70 | ||
XM_021242388.1 | XP_021098047.1 | 3 - 4 | 201 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
