Assay Details
Gene Symbol: Cers4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004865394.2 | XP_004865451.1 | 9 - 10 | 985 | 64 | |
XM_021245243.1 | XP_021100902.1 | 10 - 11 | 1078 | 64 | ||
XM_021245244.1 | XP_021100903.1 | 9 - 10 | 1906 | 64 | ||
XM_021245245.1 | XP_021100904.1 | 9 - 10 | 961 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
