Assay Details
| Gene Symbol: Lhpp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838752.3 | XP_004838809.1 | 4 - 5 | 661 | 66 | |
| XM_004838754.3 | XP_004838811.1 | 4 - 5 | 657 | 66 | ||
| XM_021253302.1 | XP_021108961.1 | 4 - 5 | 665 | 66 | ||
| XM_021253303.1 | XP_021108962.1 | 4 - 5 | 665 | 66 | ||
More Information
Set Membership: |
|

Lhpp
Genomic Map