Assay Details
Gene Symbol: LOC101700274 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004846854.2 | XP_004846911.1 | 2 - 3 | 491 | 100 | |
XM_021260041.1 | XP_021115700.1 | 3 - 4 | 1165 | 100 | ||
XM_021260042.1 | XP_021115701.1 | 3 - 4 | 1015 | 100 | ||
XM_021260043.1 | XP_021115702.1 | 4 - 5 | 798 | 100 | ||
XM_021260044.1 | XP_021115703.1 | 4 - 5 | 734 | 100 | ||
XM_021260045.1 | XP_021115704.1 | 4 - 5 | 660 | 100 | ||
XM_021260046.1 | XP_021115705.1 | 3 - 4 | 626 | 100 | ||
XM_021260047.1 | XP_021115706.1 | 2 - 3 | 505 | 100 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
