Assay Details
Gene Symbol: LOC101700274 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004846854.2 | XP_004846911.1 | 3 - 4 | 775 | 84 | |
XM_021260042.1 | XP_021115701.1 | 4 - 5 | 1299 | 84 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
