Assay Details
Gene Symbol: Tlr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021265056.1 | XP_021120715.1 | 2 - 3 | 807 | 126 | |
XM_021265057.1 | XP_021120716.1 | 3 - 4 | 585 | 126 | ||
XM_021265058.1 | XP_021120717.1 | 3 - 4 | 578 | 126 | ||
XM_021265059.1 | XP_021120718.1 | 4 - 5 | 660 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() |
