Assay Details
Gene Symbol: CUNH3orf38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004857913.3 | XP_004857970.1 | 1 - 2 | 292 | 97 | |
XM_004857914.3 | XP_004857971.1 | 1 - 2 | 292 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
