Assay Details
| Gene Symbol: Znf76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013072873.2 | XP_012928327.1 | 3 - 4 | 426 | 58 | |
| XM_013072874.2 | XP_012928328.1 | 2 - 3 | 269 | 58 | ||
| XM_021259958.1 | XP_021115617.1 | 2 - 3 | 279 | 58 | ||
| XM_021259959.1 | XP_021115618.1 | 2 - 3 | 350 | 58 | ||
| XM_021259960.1 | XP_021115619.1 | 3 - 4 | 426 | 58 | ||
More Information
Set Membership: |
|

Znf76
Genomic Map