Assay Details
Gene Symbol: Znf800 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004856412.2 | XP_004856469.2 | 4 - 5 | 959 | 86 | |
XM_021237545.1 | XP_021093204.1 | 4 - 5 | 962 | 86 | ||
XM_021237546.1 | XP_021093205.1 | 4 - 5 | 960 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
