Assay Details
| Gene Symbol: Pgm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838191.3 | XP_004838248.1 | 7 - 8 | 1389 | 61 | |
| XM_021252747.1 | XP_021108406.1 | 7 - 8 | 1389 | 61 | ||
More Information
Set Membership: |
|

Pgm5
Genomic Map