Assay Details
Gene Symbol: Nlrp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021243293.1 | XP_021098952.1 | 9 - 10 | 2997 | 67 | |
XM_021243294.1 | XP_021098953.1 | 9 - 10 | 2997 | 67 | ||
XM_021243295.1 | XP_021098954.1 | 8 - 9 | 2717 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
