Assay Details
Gene Symbol: Smpd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004842968.2 | XP_004843025.1 | 7 - 8 | 2204 | 54 | |
XM_004842970.2 | XP_004843027.1 | 6 - 7 | 2137 | 54 | ||
XM_021256803.1 | XP_021112462.1 | 7 - 8 | 2131 | 54 | ||
XM_021256804.1 | XP_021112463.1 | 7 - 8 | 2076 | 54 | ||
XM_021256805.1 | XP_021112464.1 | 7 - 8 | 2092 | 54 | ||
XM_021256806.1 | XP_021112465.1 | 7 - 8 | 2020 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
