Assay Details
| Gene Symbol: Dym | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855125.2 | XP_004855182.1 | 8 - 9 | 1091 | 63 | |
| XM_013075940.2 | XP_012931394.1 | 8 - 9 | 1091 | 63 | ||
| XM_021236605.1 | XP_021092264.1 | 8 - 9 | 1091 | 63 | ||
| XM_021236606.1 | XP_021092265.1 | 8 - 9 | 1091 | 63 | ||
| XM_021236607.1 | XP_021092266.1 | 7 - 8 | 688 | 63 | ||
More Information
Set Membership: |
|

Dym
Genomic Map