Assay Details
Gene Symbol: Riiad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004854094.3 | XP_004854151.1 | 1 - 2 | 110 | 64 | |
XM_013075450.2 | XP_012930904.1 | 1 - 2 | 110 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
