Assay Details
Gene Symbol: Fmr1nb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013067898.2 | XP_012923352.2 | 1 - 2 | 416 | 108 | |
XM_021249162.1 | XP_021104821.1 | 1 - 2 | 417 | 108 | ||
XM_021249163.1 | XP_021104822.1 | 1 - 2 | 416 | 108 | ||
XM_021249164.1 | XP_021104823.1 | 1 - 2 | 417 | 108 | ||
XM_021249165.1 | XP_021104824.1 | 1 - 2 | 415 | 108 | ||
XM_021249166.1 | XP_021104825.1 | 1 - 2 | 417 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
