Assay Details
| Gene Symbol: Nedd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847694.2 | XP_004847751.1 | 5 - 6 | 2062 | 69 | |
| XM_021260652.1 | XP_021116311.1 | 6 - 7 | 2193 | 69 | ||
| XM_021260653.1 | XP_021116312.1 | 4 - 5 | 1960 | 69 | ||
| XM_021260654.1 | XP_021116313.1 | 5 - 6 | 1913 | 69 | ||
More Information
Set Membership: |
|

Nedd9
Genomic Map