Assay Details
Gene Symbol: LOC101714047 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004849170.3 | XP_004849227.2 | 3 - 4 | 361 | 108 | |
XM_004849171.3 | XP_004849228.1 | 4 - 5 | 695 | 108 | ||
XM_004849174.3 | XP_004849231.1 | 3 - 4 | 384 | 108 | ||
XM_004849175.3 | XP_004849232.1 | 3 - 4 | 360 | 108 | ||
XM_013073651.2 | XP_012929105.1 | 4 - 5 | 435 | 108 | ||
XM_021261904.1 | XP_021117563.1 | 4 - 5 | 1529 | 108 | ||
XM_021261905.1 | XP_021117564.1 | 5 - 6 | 856 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
