Assay Details
| Gene Symbol: Supt3h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846432.2 | XP_004846489.1 | 9 - 10 | 1044 | 84 | |
| XM_004846433.2 | XP_004846490.1 | 9 - 10 | 1040 | 84 | ||
| XM_004846434.2 | XP_004846491.1 | 9 - 10 | 1033 | 84 | ||
| XM_004846435.2 | XP_004846492.1 | 8 - 9 | 948 | 84 | ||
| XM_004846436.2 | XP_004846493.1 | 7 - 8 | 689 | 84 | ||
| XM_021259730.1 | XP_021115389.1 | 10 - 11 | 1145 | 84 | ||
| XM_021259731.1 | XP_021115390.1 | 8 - 9 | 710 | 84 | ||
| XM_021259732.1 | XP_021115391.1 | 6 - 7 | 663 | 84 | ||
| XM_021259734.1 | XP_021115393.1 | 6 - 7 | 612 | 84 | ||
| XM_021259735.1 | XP_021115394.1 | 9 - 10 | 1048 | 84 | ||
| XM_021259736.1 | XP_021115395.1 | 9 - 10 | 1048 | 84 | ||
| XM_021259737.1 | XP_021115396.1 | 9 - 10 | 1051 | 84 | ||
| XM_021259738.1 | XP_021115397.1 | 6 - 7 | 631 | 84 | ||
More Information
Set Membership: |
|

Supt3h
Genomic Map