Assay Details
Gene Symbol: LOC101706848 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004869315.2 | XP_004869372.1 | 2 - 3 | 416 | 96 | |
XM_004869316.2 | XP_004869373.1 | 2 - 3 | 416 | 96 | ||
XM_004869317.3 | XP_004869374.1 | 2 - 3 | 416 | 96 | ||
XM_013067259.2 | XP_012922713.1 | 2 - 3 | 416 | 96 | ||
XM_013067261.2 | XP_012922715.1 | 2 - 3 | 307 | 96 | ||
XM_021247951.1 | XP_021103610.1 | 2 - 3 | 416 | 96 | ||
XM_021247952.1 | XP_021103611.1 | 2 - 3 | 215 | 96 | ||
XM_021247953.1 | XP_021103612.1 | 2 - 3 | 296 | 96 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
