Assay Details
| Gene Symbol: LOC101706848 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004869315.2 | XP_004869372.1 | 3 - 4 | 461 | 69 | |
| XM_004869316.2 | XP_004869373.1 | 3 - 4 | 461 | 69 | ||
| XM_004869317.3 | XP_004869374.1 | 3 - 4 | 461 | 69 | ||
| XM_013067259.2 | XP_012922713.1 | 3 - 4 | 461 | 69 | ||
| XM_013067261.2 | XP_012922715.1 | 3 - 4 | 352 | 69 | ||
| XM_021247951.1 | XP_021103610.1 | 3 - 4 | 461 | 69 | ||
| XM_021247952.1 | XP_021103611.1 | 3 - 4 | 260 | 69 | ||
| XM_021247953.1 | XP_021103612.1 | 3 - 4 | 341 | 69 | ||
More Information
Set Membership: |
|

LOC101706848
Genomic Map