Assay Details
Gene Symbol: Kiaa0513 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004842729.2 | XP_004842786.1 | 8 - 9 | 1143 | 53 | |
XM_004842730.2 | XP_004842787.1 | 8 - 9 | 1143 | 53 | ||
XM_021256617.1 | XP_021112276.1 | 8 - 9 | 1143 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
