Assay Details
Gene Symbol: Gnl3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013068785.2 | XP_012924239.1 | 5 - 6 | 399 | 60 | |
XM_021251211.1 | XP_021106870.1 | 5 - 6 | 401 | 60 | ||
XM_021251212.1 | XP_021106871.1 | 5 - 6 | 401 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
