Assay Details
Gene Symbol: Tph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004851516.2 | XP_004851573.1 | 8 - 9 | 2313 | 65 | |
XM_021263893.1 | XP_021119552.1 | 10 - 11 | 1515 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
