Assay Details
Gene Symbol: Sec14l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004843443.3 | XP_004843500.2 | 5 - 6 | 544 | 70 | |
XM_004843444.3 | XP_004843501.1 | 5 - 6 | 536 | 70 | ||
XM_013071659.2 | XP_012927113.1 | 5 - 6 | 538 | 70 | ||
XM_013071660.2 | XP_012927114.1 | 6 - 7 | 757 | 70 | ||
XM_013071661.2 | XP_012927115.1 | 5 - 6 | 537 | 70 | ||
XM_013071662.2 | XP_012927116.1 | 5 - 6 | 555 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
