Assay Details
Gene Symbol: LOC101702684 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013076264.2 | XP_012931718.1 | 2 - 3 | 484 | 122 | |
XM_013076267.2 | XP_012931721.1 | 35 - 36 | 5173 | 122 |

More Information
Set Membership: |
![]() ![]() |
