Assay Details
| Gene Symbol: Htr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854372.2 | XP_004854429.1 | 3 - 4 | 594 | 75 | |
| XM_021266178.1 | XP_021121837.1 | 3 - 4 | 435 | 75 | ||
| XM_021266179.1 | XP_021121838.1 | 3 - 4 | 314 | 75 | ||
| XM_021266180.1 | XP_021121839.1 | 3 - 4 | 435 | 75 | ||
| XM_021266181.1 | XP_021121840.1 | 3 - 4 | 435 | 75 | ||
More Information
Set Membership: |
|

Htr4
Genomic Map