Assay Details
| Gene Symbol: Gnb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004869451.3 | XP_004869508.1 | 9 - 10 | 1819 | 73 | |
| XM_013067324.2 | XP_012922778.1 | 7 - 8 | 1693 | 73 | ||
| XM_021248028.1 | XP_021103687.1 | 9 - 10 | 1792 | 73 | ||
| XM_021248029.1 | XP_021103688.1 | 8 - 9 | 1645 | 73 | ||
More Information
Set Membership: |
|

Gnb3
Genomic Map