Assay Details
| Gene Symbol: Zar1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854765.2 | XP_004854822.1 | 1 - 2 | 1939 | 97 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zar1l| Gene Symbol: Zar1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854765.2 | XP_004854822.1 | 1 - 2 | 1939 | 97 | |