Assay Details
| Gene Symbol: Zar1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854765.2 | XP_004854822.1 | 2 - 3 | 2043 | 104 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zar1l| Gene Symbol: Zar1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854765.2 | XP_004854822.1 | 2 - 3 | 2043 | 104 | |
Set Membership: |
|