Assay Details
Gene Symbol: Stx16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004840941.3 | XP_004840998.1 | 4 - 5 | 1191 | 66 | |
XM_004840942.3 | XP_004840999.1 | 3 - 4 | 1180 | 66 | ||
XM_004840943.3 | XP_004841000.1 | 4 - 5 | 1184 | 66 | ||
XM_004840944.3 | XP_004841001.1 | 3 - 4 | 1172 | 66 | ||
XM_004840945.3 | XP_004841002.1 | 3 - 4 | 1408 | 66 | ||
XM_004840946.3 | XP_004841003.1 | 4 - 5 | 1138 | 66 | ||
XM_004840947.3 | XP_004841004.1 | 3 - 4 | 1126 | 66 | ||
XM_004840948.3 | XP_004841005.1 | 4 - 5 | 1131 | 66 | ||
XM_004840949.3 | XP_004841006.1 | 3 - 4 | 1119 | 66 | ||
XM_004840951.3 | XP_004841008.1 | 4 - 5 | 1186 | 66 | ||
XM_013070790.2 | XP_012926244.1 | 4 - 5 | 1186 | 66 | ||
XM_021255166.1 | XP_021110825.1 | 5 - 6 | 1308 | 66 | ||
XM_021255167.1 | XP_021110826.1 | 5 - 6 | 1257 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
