Assay Details
| Gene Symbol: Washc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021245278.1 | XP_021100937.1 | 8 - 9 | 1253 | 74 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Washc1| Gene Symbol: Washc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021245278.1 | XP_021100937.1 | 8 - 9 | 1253 | 74 | |