Assay Details
| Gene Symbol: Washc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865313.3 | XP_004865370.1 | 9 - 10 | 1422 | 88 | |
| XM_004865314.2 | XP_004865371.1 | 9 - 10 | 1422 | 88 | ||
| XM_021245270.1 | XP_021100929.1 | 9 - 10 | 1423 | 88 | ||
| XM_021245271.1 | XP_021100930.1 | 11 - 12 | 1695 | 88 | ||
| XM_021245272.1 | XP_021100931.1 | 9 - 10 | 1419 | 88 | ||
| XM_021245273.1 | XP_021100932.1 | 9 - 10 | 1285 | 88 | ||
| XM_021245274.1 | XP_021100933.1 | 9 - 10 | 1325 | 88 | ||
| XM_021245275.1 | XP_021100934.1 | 9 - 10 | 1465 | 88 | ||
| XM_021245277.1 | XP_021100936.1 | 9 - 10 | 2413 | 88 | ||
| XM_021245278.1 | XP_021100937.1 | 9 - 10 | 1419 | 88 | ||
| XM_021245279.1 | XP_021100938.1 | 9 - 10 | 1399 | 88 | ||
More Information
Set Membership: |
|

Washc1
Genomic Map