Assay Details
| Gene Symbol: CUNH14orf180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836867.3 | XP_004836924.1 | 1 - 2 | 356 | 88 | |
| XM_021243642.1 | XP_021099301.1 | 2 - 3 | 232 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004836867.3, XM_021243642.1 transcripts.
Set Membership: |
|

CUNH14orf180
Genomic Map