Assay Details
| Gene Symbol: CUNH14orf180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836867.3 | XP_004836924.1 | 4 - 5 | 649 | 93 | |
| XM_013066255.2 | XP_012921709.1 | 5 - 6 | 728 | 93 | ||
| XM_021243642.1 | XP_021099301.1 | 5 - 6 | 525 | 93 | ||

CUNH14orf180
Genomic Map