Assay Details
Gene Symbol: Itgb3bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004841524.3 | XP_004841581.1 | 3 - 4 | 346 | 90 | |
XM_021255622.1 | XP_021111281.1 | 3 - 4 | 285 | 90 | ||
XM_021255623.1 | XP_021111282.1 | 3 - 4 | 259 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
